missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Sorbitol-1,1,2,3,4,5,6,6-d8, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | D-Glucitol-d8 |
| CAS | 287962-59-8 |
| Molecular Formula | C6 D8 H6 O6 |
| Quantity | 0.05 g |
| Synonym | Cordycepic Acid-d8, Brightmoon-d8, Bronchitol-d8, D-(-)-Mannitol-d8, D-Mannitol-d8, D-Mannit-d8, Diosmol-d8, Isotol-d8, Manicol-d8, Maniton S-d8, Mannidex-d8, Mannigen-d8, Mannistol-d8, Mannit-d8, Mannit 60-d8, Mannit P-d8, Mannit S-d8, Mannite-d8, Mannitol-d8, Mannitol 35-d8, Mannitol 60-d8, Mannitol P 60-d8, Mannitolum-d8, Mannogem 2080-d8, Mannogem EZ-d8, Marine Crystal-d8, Nonpareil 108-d8, Orocell 200-d8, Osmitrol-d8, Osmosal-d8, Parteck 300-d8, Parteck Delta M-d8, Parteck M 100-d8, Parteck M 200-d8, Parteck M 300-d8, Partek M-d8, Pearlitol-d8, Perteck M-d8, Resectisol-d8, D-Sorbitol-d8, Glucarinev-d8, Esasorb-d8, Cholaxine-d8, Karion-d8, Sionite-d8, Sionon-d8 |
| InChI Formula | InChI=1 S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12 H,1-2H2/t3-,4-,5-,6-/m1/s1/i1D2,2D2,3 D,4 D,5 D,6 D |
| SMILES | [2 H]C([2 H])(O)[C@@]([2 H])(O)[C@@]([2 H])(O)[C@]([2 H])(O)[C@]([2 H])(O)C([2 H])([2 H])O |
| IUPAC Name | (2 R,3 R,4 R,5 R)-1,1,2,3,4,5,6,6-octadeuteriohexane-1,2,3,4,5,6-hexol |
| Molecular Weight (g/mol) | 190.22 |
| Formula Weight | 190.1293 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction