missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatinine-d5 (N-methyl-d3; imidazolidinone-5,5-d2), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Creatinine-D5 |
| Molecular Formula | C4 D5 H2 N3 O |
| Quantity | 0.05 g |
| Synonym | 2-Amino-1,5-dihydro-1-(methyl-d3)-4 H-imidazol-4-one-5,5-d2, 2-Amino-3-(trideuteriomethyl)-5,5-dideuterio-4 H-imidazol-5-one, Creatinine-d5, Creatinine-D5, Creatinine-d5 (N-methyl-d3, imidazolidinone-5,5-d2) |
| InChI Formula | InChI=1 S/C4H7N3O/c1-7-2-3(8)6-4(7)5/h2H2,1H3,(H2,5,6,8)/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])N1C(=NC(=O)C1([2 H])[2 H])N |
| IUPAC Name | 2-amino-5,5-dideuterio-1-(trideuteriomethyl)imidazol-4-one |
| Molecular Weight (g/mol) | 118.1487 |
| Formula Weight | 118.0903 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction