missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(3-aminopropyl-d6)amine, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 3,3'-Diaminodipropylamine-D12 |
| CAS | 1219805-55-6 |
| Molecular Formula | C6 D12 H5 N3 |
| Quantity | 0.05 g |
| Synonym | Bis(3-aminopropyl-d6)amine, Bis(3-aminopropyl)amine-d12, 3,3'-Diaminodipropylamine-D12, 3,3'-Diaminodipropylamine-d12, N-(3-Aminopropyl-1,1,2,2,3,3-d6)-1,3-propanediamine-1,1,2,2,3,3-d6 |
| InChI Formula | InChI=1 S/C6H17N3/c7-3-1-5-9-6-2-4-8/h9H,1-8H2/i1D2,2D2,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])(N)C([2 H])([2 H])C([2 H])([2 H])NC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N |
| IUPAC Name | N'-(3-amino-1,1,2,2,3,3-hexadeuteriopropyl)-1,1,2,2,3,3-hexadeuteriopropane-1,3-diamine |
| Molecular Weight (g/mol) | 143.2932 |
| Formula Weight | 143.2176 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction