Learn More
Bis(2-hydroxyethyl)-1,1,2,2-d4-amine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Diethanolamine-D4 (2-hydroxyethyl-1,1,2,2-D4) |
| CAS | 352431-07-3 |
| Molecular Formula | C4 D4 H7 N O2 |
| Quantity | 0.05 g |
| Synonym | Ethan-1,1,2,2-d4-ol, 2-[(2-hydroxyethyl)amino]- (9 CI, ACI), 2-[(2-Hydroxyethyl)amino]ethan-1,1,2,2-d4-ol (ACI), Bis(2-hydroxyethyl)-1,1,2,2-d4-amine, Bis(2-hydroxyethyl-1,1,2,2-d4)amine, Diethanolamine-d4, Diethanolamine-D4 (2-hydroxyethyl-1,1,2,2-D4) |
| InChI Formula | InChI=1 S/C4H11NO2/c6-3-1-5-2-4-7/h5-7 H,1-4H2/i1D2,3D2 |
| SMILES | [2 H]C([2 H])(O)C([2 H])([2 H])NCCO |
| IUPAC Name | 1,1,2,2-tetradeuterio-2-(2-hydroxyethylamino)ethanol |
| Molecular Weight (g/mol) | 109.1603 |
| Formula Weight | 109.1041 g/mol |
| Show More |