Learn More
Bis[(±)-2-ethylhexyl] Phthalate-d38, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Bis(2-ethylhexyl) Phthalate-D38 |
| CAS | 352431-42-6 |
| Molecular Formula | C24 D38 O4 |
| Quantity | 0.05 g |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, bis[2-(ethyl-d5)hexyl-1,1,2,3,3,4,4,5,5,6,6,6-d12] ester (9 CI), 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid bis[2-(ethyl-d5)hexyl-1,1,2,3,3,4,4,5,5,6,6,6-d12] ester, Bis[(±)-2-ethylhexyl] Phthalate-d38, Diethylhexylphthalate-d38, Bis(2-ethylhexyl) Phthalate-D38, Phthalic Acid Bis(2-ethylhexyl) Ester D38 |
| InChI Formula | InChI=1 S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20 H,5-10,13-14,17-18H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11 D,12 D,13D2,14D2,15 D,16 D,17D2,18D2,19 D,20 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OC([2 H])([2 H])C([2 H])(C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])c(C(=O)OC([2 H])([2 H])C([2 H])(C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])c1[2 H] |
| IUPAC Name | bis[1,1,2,3,3,4,4,5,5,6,6,6-dodecadeuterio-2-(1,1,2,2,2-pentadeuterioethyl)hexyl] 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 428.7903 |
| Formula Weight | 428.5155 g/mol |
| Show More |