Learn More
Benzimidazole-4,5,6,7-d4, CDN
Product Code. 30311390
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30311390
Brand: CDN CDND40790.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Benzimidazole-4,5,6,7-D4
CAS 64531-29-9
Molecular Formula C7 D4 H2 N2
Quantity 0.05 g
Synonym 1 H-Benzimidazole-4,5,6,7-d4 (9 CI), Benzimidazole-4,5,6,7-d4, Benzimidazole-d4, Benzimidazole-4,5,6,7-D4
InChI Formula InChI=1 S/C7H6N2/c1-2-4-7-6(3-1)8-5-9-7/h1-5 H,(H,8,9)/i1D,2 D,3 D,4 D
SMILES [2 H]c1c([2 H])c([2 H])c2[nH]cnc2c1[2 H]
IUPAC Name 4,5,6,7-tetradeuterio-1 H-benzimidazole
Molecular Weight (g/mol) 122.1606
Formula Weight 122.0782 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity