Learn More
Acetyl-d3-L-carnitine HCl, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Acetyl-d3 L-Carnitine Hydrochloride |
| CAS | 362049-62-5 |
| Molecular Formula | C9 2H3 H15 N O4 . Cl |
| Quantity | 0.05 g |
| Synonym | 1-Propanaminium, 2-(acetyl-d3-oxy)-3-carboxy-N,N,N-trimethyl-, chloride, (2 R)- (9 CI), Acetyl-d3 L-Carnitine Chloride, Acetyl-d3 L-Carnitine Hydrochloride |
| InChI Formula | InChI=1 S/C9H17NO4.ClH/c1-7(11)14-8(5-9(12)13)6-10(2,3)4;/h8H,5-6H2,1-4H3;1 H/t8-;/m1./s1/i1D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])C(=O)O[C@H](CC(=O)O)C[N+](C)(C)C |
| IUPAC Name | [(2 R)-3-carboxy-2-(2,2,2-trideuterioacetyl)oxypropyl]-trimethylazanium;chloride |
| Molecular Weight (g/mol) | 242.72 |
| Formula Weight | 242.1113 g/mol |
| Show More |