missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Cholesten-24(RS)-methyl-3beta-ol-25,26,26,26,27,27,27-d7, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Campesterol-d7 (Mixture of Diastereomers) |
| CAS | 2483832-11-5 |
| Molecular Formula | C28 D7 H41 O |
| Quantity | 0.05 g |
| Synonym | Ergost-5-en-25,26,26,26,27,27,27-d7-3-ol, (3β,24ξ)- (ACI), (3β,24ξ)-Ergost-5-en-25,26,26,26,27,27,27-d7-3-ol (ACI), (24ξ)-24-Methylcholest-5-en-3β-ol-25,26,26,26,27,27,27-d7, (3β,24ξ)-Ergost-5-en-3-ol-25,26,26,26,27,27,27-D7, 5-Cholesten-24(RS)-methyl-3β-ol-25,26,26,26,27,27,27-d7 |
| InChI Formula | InChI=1 S/C28H48O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h9,18-20,22-26,29 H,7-8,10-17H2,1-6H3/t19?,20-,22+,23+,24-,25+,26+,27+,28-/m1/s1/i1D3,2D3,18 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C(C)CC[C@@H](C)[C@H]1 CC[C@H]2 [C@@H]3 CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C)C([2 H])([2 H])[2 H] |
| IUPAC Name | (3 S,8 S,9 S,10 R,13 R,14 S,17 R)-10,13-dimethyl-17-[(2 R)-6,7,7,7-tetradeuterio-5-methyl-6-(trideuteriomethyl)heptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-3-ol |
| Molecular Weight (g/mol) | 407.7233 |
| Formula Weight | 407.4145 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction