Learn More
4-n-Pentylphenol-d16, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Pentylphenol-D16 |
| CAS | 1219805-40-9 |
| Molecular Formula | C11 D16 O |
| Quantity | 0.05 g |
| Synonym | 4-Pentylphenol-D16, 4-n-Pentylphenol-d16, 4-n-Amylphenol D16, 4-(Pentyl-1,1,2,2,3,3,4,4,5,5,5-d11)phen-2,3,5,6-d4-ol-d |
| InChI Formula | InChI=1 S/C11H16O/c1-2-3-4-5-10-6-8-11(12)9-7-10/h6-9,12 H,2-5H2,1H3/i1D3,2D2,3D2,4D2,5D2,6 D,7 D,8 D,9 D/hD |
| SMILES | [2 H]Oc1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,4,5-tetradeuterio-3-deuteriooxy-6-(1,1,2,2,3,3,4,4,5,5,5-undecadeuteriopentyl)benzene |
| Molecular Weight (g/mol) | 180.3427 |
| Formula Weight | 180.2205 g/mol |
| Show More |