missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(N-Methyl-N-nitrosoamino)-1-(3-pyridyl-d4)-1-butanone, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | N-Nitrosonornicotine-ketoneD4(pyridylD4) |
| CAS | 764661-24-7 |
| Molecular Formula | C10 2H4 H9 N3 O2 |
| Quantity | 0.05 g |
| Synonym | 1-Butanone, 4-(methylnitrosoamino)-1-(3-pyridinyl-2,4,5,6-d4)-, N-Nitrosonornicotine-ketone D4 (pyridyl D4), 4-(Methylnitrosamino)-1-(3-pyridyl-d4)-1-butanone |
| InChI Formula | InChI=1 S/C10H13N3O2/c1-13(12-15)7-3-5-10(14)9-4-2-6-11-8-9/h2,4,6,8 H,3,5,7H2,1H3/i2D,4 D,6 D,8 D |
| SMILES | [2 H]c1nc([2 H])c(C(=O)CCCN(C)N=O)c([2 H])c1[2 H] |
| IUPAC Name | N-methyl-N-[4-oxo-4-(2,4,5,6-tetradeuteriopyridin-3-yl)butyl]nitrous amide |
| Molecular Weight (g/mol) | 211.25 |
| Formula Weight | 211.1259 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction