missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(n-Butylamino-2,2,3,3,4,4,4-d7)benzoic Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-(Butylamino)benzoic acid butyl-2,2,3,3,4,4,4-d7 |
| CAS | 1219803-44-7 |
| Molecular Formula | C11 2H7 H8 N O2 |
| Quantity | 0.05 g |
| Synonym | 4-(Butylamino)benzoic Acid D7 (butyl-2,2,3,3,4,4,4-D7), 4-(n-Butylamino-2,2,3,3,4,4,4-d7)benzoic acid |
| InChI Formula | InChI=1 S/C11H15NO2/c1-2-3-8-12-10-6-4-9(5-7-10)11(13)14/h4-7,12 H,2-3,8H2,1H3,(H,13,14)/i1D3,2D2,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CNc1ccc(cc1)C(=O)O |
| IUPAC Name | 4-(2,2,3,3,4,4,4-heptadeuteriobutylamino)benzoic acid |
| Molecular Weight (g/mol) | 200.29 |
| Formula Weight | 200.1542 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction