Learn More
4-Cholesten-3-one-4-13C, CDN
Click to view available options
Quantity:
0.05 g
Unit Size:
0.05g
Specifications
Specifications
| Chemical Name or Material | 4-Cholesten-3-one-4-13 C |
| CAS | 68165-57-1 |
| Molecular Formula | 13 C C26 H44 O |
| Quantity | 0.05 g |
| Synonym | Cholest-4-en-3-one-4-13 C (9 CI), 4-Cholesten-3-one-4-13 C, delta;4-Cholesten-3-one-4-13 C, (17β)-17-Octylandrost-4-en-3-one-4-13 C |
| InChI Formula | InChI=1 S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h17-19,22-25 H,6-16H2,1-5H3/t19-,22+,23-,24+,25+,26+,27-/m1/s1/i17+1 |
| SMILES | CC(C)CCC[C@@H](C)[C@H]1 CC[C@H]2 [C@@H]3 CCC4=[13 CH]C(=O)CC[C@]4(C)[C@H]3 CC[C@]12 C |
| IUPAC Name | (8 S,9 S,10 R,13 R,14 S,17 R)-10,13-dimethyl-17-[(2 R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 385.6303 |
| Formula Weight | 385.3426 g/mol |
| Show More |