Learn More
4-Bromo-alpha,alpha,alpha-trifluorotoluene-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Bromobenzotrifluoride-D4 |
| CAS | 1219799-09-3 |
| Molecular Formula | C7 D4 Br F3 |
| Quantity | 0.05 g |
| Synonym | 4-Bromo-α,α,α-trifluorotoluene-d4, 4-Bromobenzotrifluoride-d4, 4-Bromobenzotrifluoride-D4, 1-Bromo-4-(trifluoromethyl)benzene-2,3,5,6-d4, 3-Bromo-6-(trifluoromethyl)benzene-1,2,4,5-d |
| InChI Formula | InChI=1 S/C7H4BrF3/c8-6-3-1-5(2-4-6)7(9,10)11/h1-4 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1Br)C(F)(F)F |
| IUPAC Name | 1-bromo-2,3,5,6-tetradeuterio-4-(trifluoromethyl)benzene |
| Molecular Weight (g/mol) | 229.0305 |
| Formula Weight | 227.97 g/mol |
| Show More |