missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-d3-indole, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 3-Methyl-d3-indole |
| CAS | 111399-60-1 |
| Molecular Formula | C9 2H3 H6 N |
| Quantity | 0.05 g |
| Synonym | 1 H-Indole, 3-(methyl-d3)-, 3-(Methyl-d3)-1 H-indole, 3-Methyl-1 H-indole-d3, 3-Methylindole-D3 (methyl-D3), Scatole-d3, Skatol-d3, β-Methylindole-d3 |
| InChI Formula | InChI=1 S/C9H9N/c1-7-6-10-9-5-3-2-4-8(7)9/h2-6,10 H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])c1c[nH]c2ccccc12 |
| IUPAC Name | 3-(trideuteriomethyl)-1 H-indole |
| Molecular Weight (g/mol) | 134.19 |
| Formula Weight | 134.0923 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction