Learn More
(3,4-Dihydroxyphenyl-d3)acetic-2,2-d2 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 3,4-Dihydroxyphenylacetic Acid-d5 |
| CAS | 60696-39-1 |
| Molecular Formula | C8 D5 H3 O4 |
| Quantity | 0.05 g |
| Synonym | Benzene-2,3,6-d3-acetic-α,α-d2 acid, 4,5-dihydroxy- (9 CI), 3,4-Dihydroxybenzene-2,5,6-d3-acetic-α,α-d2 acid, (3,4-Dihydroxyphenyl-2,5,6-d3)acetic-d2 acid, 2-(3,4-Dihydroxyphenyl-2,5,6-d3)acetic-2,2-d2 acid, 3',4'-Dihydroxyphenylacetic-d5 acid, 3',4'-Dihydroxyphenylacetic acid-2,2,2',5',6'-d5, 3',4'-Dihydroxyphenylacetic acid-2,2,2',5',6'-d5 |
| InChI Formula | InChI=1 S/C8H8O4/c9-6-2-1-5(3-7(6)10)4-8(11)12/h1-3,9-10 H,4H2,(H,11,12)/i1D,2 D,3 D,4D2 |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c(O)c1O)C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2-dideuterio-2-(2,3,6-trideuterio-4,5-dihydroxyphenyl)acetic acid |
| Molecular Weight (g/mol) | 173.178 |
| Formula Weight | 173.074 g/mol |
| Show More |