missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3',5,5'-Tetrabromobisphenol A-2,2',6,6'-d4, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Tetrabromobisphenol A-d6 |
| Molecular Formula | C15 D4 H8 Br4 O2 |
| Quantity | 0.05 g |
| Synonym | 3,3',5,5'-Tetrabromobisphenol A D4 (ring D4), 3,3',5,5'-Tetrabromobisphenol A-2,2',6,6'-d4 |
| InChI Formula | InChI=1 S/C15H12Br4O2/c1-15(2,7-3-9(16)13(20)10(17)4-7)8-5-11(18)14(21)12(19)6-8/h3-6,20-21 H,1-2H3/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c(Br)c(O)c(Br)c([2 H])c1C(C)(C)c2c([2 H])c(Br)c(O)c(Br)c2[2 H] |
| IUPAC Name | 2,6-dibromo-3,5-dideuterio-4-[2-(3,5-dibromo-2,6-dideuterio-4-hydroxyphenyl)propan-2-yl]phenol |
| Molecular Weight (g/mol) | 547.8952 |
| Formula Weight | 543.7822 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction