Learn More
2-Methylfuran-d6, CDN
Product Code. 30324518
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30324518
Brand: CDN CDND75120.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methylfuran-d6
CAS 1398065-93-4
Molecular Formula C5D6O
Quantity 0.05 g
Synonym 5-Methylfuran-d6, NSC 3707-d6, NSC 5211-d6, Silvan-d6, Sylvan-d6, α-Methylfuran-d6;
InChI Formula InChI=1 S/C5H6O/c1-5-3-2-4-6-5/h2-4 H,1H3/i1D3,2 D,3 D,4 D
SMILES [2 H]c1oc(c([2 H])c1[2 H])C([2 H])([2 H])[2 H]
IUPAC Name 2,3,4-trideuterio-5-(trideuteriomethyl)furan
Molecular Weight (g/mol) 88.14
Formula Weight 88.0795 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity