Learn More
2-Aminofluorene-d11, CDN
Product Code. 30319654
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319654
Brand: CDN CDND38040.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Aminofluorene-D11
CAS 347841-44-5
Molecular Formula C13 D11 N
Quantity 0.05 g
Synonym 9 H-Fluoren-1,3,4,5,6,7,8,9,9-d9-2-amine-d2 (9 CI), 2-Aminofluorene-d11, 2-Aminofluorene-D11
InChI Formula InChI=1 S/C13H11N/c14-11-5-6-13-10(8-11)7-9-3-1-2-4-12(9)13/h1-6,8 H,7,14H2/i1D,2 D,3 D,4 D,5 D,6 D,7D2,8 D/hD2
SMILES [2 H]N([2 H])c1c([2 H])c([2 H])c2c3c([2 H])c([2 H])c([2 H])c([2 H])c3C([2 H])([2 H])c2c1[2 H]
IUPAC Name N,N,1,3,4,5,6,7,8,9,9-undecadeuteriofluoren-2-amine
Molecular Weight (g/mol) 192.3009
Formula Weight 192.1582 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity