missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-iso-butyric-d6 Acid (dimethyl-d6), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2-Methylalanine dimethyl-d6 |
| CAS | 50348-93-1 |
| Molecular Formula | C4 2H6 H3 N O2 . Cl H |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C4H9NO2.ClH/c1-4(2,5)3(6)7;/h5H2,1-2H3,(H,6,7);1 H/i1D3,2D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C(N)(C(=O)O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-amino-3,3,3-trideuterio-2-(trideuteriomethyl)propanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 145.62 |
| Formula Weight | 145.0777 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction