Learn More
2,4-Dimethylaniline-d11, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2,4-Dimethylaniline-D11 |
| CAS | 1398065-83-2 |
| Molecular Formula | C8 D11 N |
| Quantity | 0.05 g |
| Synonym | 4,6-Di(methyl-d3)benzen-2,3,5-d3-amine-d2, 2,4-Dimethylaniline-d11, 2,4-Dimethylaniline-D11, 2,4-Xylidine-d11, 2,4-Di(methyl-d3)benzenamine-N,N,3,5,6-d5 |
| InChI Formula | InChI=1 S/C8H11N/c1-6-3-4-8(9)7(2)5-6/h3-5 H,9H2,1-2H3/i1D3,2D3,3 D,4 D,5 D/hD2 |
| SMILES | [2 H]N([2 H])c1c([2 H])c([2 H])c(c([2 H])c1C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N,2,3,5-pentadeuterio-4,6-bis(trideuteriomethyl)aniline |
| Molecular Weight (g/mol) | 132.2474 |
| Formula Weight | 132.1582 g/mol |
| Show More |