missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethyl-d6-quinoxaline, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2,3-Dimethylquinoxaline-D6 (dimethyl-D6) |
| Molecular Formula | C10 D6 H4 N2 |
| Quantity | 0.05 g |
| Synonym | 2,3-Di(methyl-d3)quinoxaline, 2,3-Dimethylquinoxaline-D6 (dimethyl-D6), 2,3-Dimethyl-d6-quinoxaline |
| InChI Formula | InChI=1 S/C10H10N2/c1-7-8(2)12-10-6-4-3-5-9(10)11-7/h3-6 H,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])c1nc2ccccc2nc1C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3-bis(trideuteriomethyl)quinoxaline |
| Molecular Weight (g/mol) | 164.2368 |
| Formula Weight | 164.1221 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction