missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethylpropionyl-L-carnitine-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Pivaloyl-L-carnitine-D3 (N-methyl-D3) Chloride |
| Molecular Formula | C12 D3 H21 N O4 . Cl |
| Quantity | 0.05 g |
| Synonym | Pivaloyl-L-carnitine-d3 chloride, Pivaloyl-L-carnitine-D3 (N-methyl-D3) Chloride, (2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-(2,2-dimethyl-1-oxopropoxy)-1-propanaminium chloride, [(2 R)-3-Carboxy-2-(2,2-dimethylpropanoyloxy)propyl]dimethyl(methyl-d3)ammonium chloride, 2,2-Dimethylpropionyl-L-carnitine-d3 HCl (N-methyl-d3) |
| InChI Formula | InChI=1 S/C12H23NO4.ClH/c1-12(2,3)11(16)17-9(7-10(14)15)8-13(4,5)6;/h9H,7-8H2,1-6H3;1 H/t9-;/m1./s1/i4D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)C(C)(C)C |
| IUPAC Name | [(2 R)-3-carboxy-2-(2,2-dimethylpropanoyloxy)propyl]-dimethyl-(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 284.7948 |
| Formula Weight | 284.1582 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction