Learn More
2,2'-Binaphthyl-d14, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2,2'-Binaphthyl-d14 |
| CAS | 210487-05-1 |
| Molecular Formula | C20 D14 |
| Quantity | 0.05 g |
| Synonym | 2,2'-Binaphthalene-1,1',3,3',4,4',5,5',6,6',7,7',8,8'-d14 (9 CI), 2,2'-Binaphthalene-d14, 2,2'-Binaphthyl-D14, 2,2'-Dinaphthyl-d14, 2-(Naphthalen-2-yl-1,3,4,5,6,7,8-d7)naphthalene-1,3,4,5,6,7,8-d7, β,β'-Binaphthyl-d14 |
| InChI Formula | InChI=1 S/C20H14/c1-3-7-17-13-19(11-9-15(17)5-1)20-12-10-16-6-2-4-8-18(16)14-20/h1-14 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D,13 D,14 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c([2 H])c(c([2 H])c([2 H])c2c1[2 H])c3c([2 H])c([2 H])c4c([2 H])c([2 H])c([2 H])c([2 H])c4c3[2 H] |
| IUPAC Name | 1,2,3,4,5,6,8-heptadeuterio-7-(1,3,4,5,6,7,8-heptadeuterionaphthalen-2-yl)naphthalene |
| Molecular Weight (g/mol) | 268.4114 |
| Formula Weight | 268.1974 g/mol |
| Show More |