Learn More
17beta-Estradiol-2,4,16,16,17-d5, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 17 beta-Estradiol-D5 |
| CAS | 221093-45-4 |
| Molecular Formula | C18 2H5 H19 O2 |
| Quantity | 0.05 g |
| Synonym | Estra-1,3,5(10)-triene-2,4,16,16,17-d5-3,17-diol, (17β)-, (17β)-Estra-1,3,5(10)-triene-2,4,16,16,17-d5-3,17-diol, 17β-Estradiol-D5, 17β-Estradiol-2,4,16,16,17-D5 |
| InChI Formula | InChI=1 S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20 H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1/i3D,7D2,10 D,17 D |
| SMILES | [2 H]c1cc2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])[C@]4([2 H])O)[C@@H]3 CCc2c([2 H])c1O |
| IUPAC Name | (8 R,9 S,13 S,14 S,17 S)-2,4,16,16,17-pentadeuterio-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 277.41 |
| Formula Weight | 277.209 g/mol |
| Show More |