missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-12-Hydroxyoctadecanoic-11,11,12,13,13-d5 Acid, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 12-Hydroxystearic Acid-11,11,12,13,13-D5 |
| CAS | 2468637-39-8 |
| Molecular Formula | C18 D5 H31 O3 |
| Quantity | 0.05 g |
| Synonym | 12-Hydroxystearic Acid-d5, (±)-12-Hydroxyoctadecanoic-11,11,12,13,13-d5 Acid, 12-Hydroxystearic Acid-11,11,12,13,13-D5 |
| InChI Formula | InChI=1 S/C18H36O3/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21/h17,19 H,2-16H2,1H3,(H,20,21)/i14D2,15D2,17 D |
| SMILES | [2 H]C([2 H])(CCCCC)C([2 H])(O)C([2 H])([2 H])CCCCCCCCCC(=O)O |
| IUPAC Name | 11,11,12,13,13-pentadeuterio-12-hydroxyoctadecanoic acid |
| Molecular Weight (g/mol) | 305.5075 |
| Formula Weight | 305.2978 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction