missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Benzene-2,3,5,6-d4-dimethanol, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1,4-Di(hydroxymethyl)benzene-d4 |
| CAS | 1158734-28-1 |
| Molecular Formula | C8 D4 H6 O2 |
| Quantity | 0.05 g |
| Synonym | 1,4-Benzene-2,3,5,6-d4-dimethanol, 1,4-Di(hydroxymethyl)benzene-d4, [2,3,5,6-Tetradeuterio-4-(hydroxymethyl)phenyl]methanol, 1,4-Di(hydroxymethyl)benzene-2,3,5,6-D4, 1,4-Phenylenedimethanol-d4, α,α'-Dihydroxy-p-xylene-d4, α,α'-Dihydroxy-p-xylene-d4 |
| InChI Formula | InChI=1 S/C8H10O2/c9-5-7-1-2-8(6-10)4-3-7/h1-4,9-10 H,5-6H2/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(CO)c([2 H])c([2 H])c1CO |
| IUPAC Name | [2,3,5,6-tetradeuterio-4-(hydroxymethyl)phenyl]methanol |
| Molecular Weight (g/mol) | 142.188 |
| Formula Weight | 142.093 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction