Learn More
1,3-Propane-d6-diamine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1,3-Diaminopropane-d6 |
| CAS | 90375-98-7 |
| Molecular Formula | C3 D6 H4 N2 |
| Quantity | 0.05 g |
| Synonym | 1,3-Propane-1,1,2,2,3,3-d6-diamine (9 CI), 1,1,2,2,3,3-Hexadeuteriopropane-1,3-diamine, 1,3-Diaminopropane-d6, 1,3-Diaminopropane-D6, Trimethylenediamine-d6, 1,3-Propane-d6-diamine |
| InChI Formula | InChI=1 S/C3H10N2/c4-2-1-3-5/h1-5H2/i1D2,2D2,3D2 |
| SMILES | [2 H]C([2 H])(N)C([2 H])([2 H])C([2 H])([2 H])N |
| IUPAC Name | 1,1,2,2,3,3-hexadeuteriopropane-1,3-diamine |
| Molecular Weight (g/mol) | 80.162 |
| Formula Weight | 80.122 g/mol |
| Show More |