Learn More
1,3-Dichloropropene-d4 (cis/trans mixture), CDN
Product Code. 30321189
Click to view available options
Quantity:
0.05 g
Unit Size:
0.05g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321189
Brand: CDN CDND26690.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,3-Dichloropropene-d4 (E,Z mixture)
CAS 202656-23-3
Molecular Formula C3 D4 Cl2
Quantity 0.05 g
Synonym cis/trans-1,3-Dichloropropene-d4
InChI Formula InChI=1 S/C3H4Cl2/c4-2-1-3-5/h1-2 H,3H2/i1D,2 D,3D2
SMILES [2 H]C(=C([2 H])C([2 H])([2 H])Cl)Cl
IUPAC Name 1,3-dichloro-1,2,3,3-tetradeuterio-prop-1-ene
Molecular Weight (g/mol) 114.99
Formula Weight 113.9941 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity