Learn More
1,10-Decane-d20-diol, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.05g
Specifications
Specifications
| Chemical Name or Material | 1,10-Decanediol-D20 |
| CAS | 347841-78-5 |
| Molecular Formula | C10 D20 H2 O2 |
| Quantity | 0.05 g |
| Synonym | 1,10-Decane-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-d20-diol (9 CI), 1,10-Decanediol-D20, 1,10-Decane-d20-diol |
| InChI Formula | InChI=1 S/C10H22O2/c11-9-7-5-3-1-2-4-6-8-10-12/h11-12 H,1-10H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2 |
| SMILES | [2 H]C([2 H])(O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-icosadeuteriodecane-1,10-diol |
| Molecular Weight (g/mol) | 194.4037 |
| Formula Weight | 194.2875 g/mol |
| Show More |