Learn More
3-Hydroxyfluoranthene-d9, CDN
Click to view available options
Quantity:
0.01 g
0.025 g
Unit Size:
0.01g
0.03g
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxyfluoranthene-D9 |
| CAS | 2733214-80-5 |
| Molecular Formula | C16 D9 H O |
| Quantity | 0.025 g |
| Synonym | 3-Fluoranthenol-d9, 3-Hydroxyfluoranthene-D9, 3-Hydroxyfluoranthene-1,2,4,5,6,7,8,9,10-d9 |
| InChI Formula | InChI=1 S/C16H10O/c17-15-9-8-13-11-5-2-1-4-10(11)12-6-3-7-14(15)16(12)13/h1-9,17 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c3c([2 H])c([2 H])c([2 H])c4c(O)c([2 H])c([2 H])c2c34 |
| IUPAC Name | 1,2,4,5,6,7,8,9,10-nonadeuteriofluoranthen-3-ol |
| Molecular Weight (g/mol) | 227.3055 |
| Formula Weight | 227.1297 g/mol |
| Show More |