missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zaleplon-d5 (N-ethyl-d5), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Zaleplon-D5 |
| CAS | 1001083-56-2 |
| Molecular Formula | C17 2H5 H10 N5 O |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C17H15N5O/c1-3-21(12(2)23)15-6-4-5-13(9-15)16-7-8-19-17-14(10-18)11-20-22(16)17/h4-9,11 H,3H2,1-2H3/i1D3,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])N(C(=O)C)c1cccc(c1)c2ccnc3c(cnn23)C#N |
| IUPAC Name | N-[3-(3-cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-(1,1,2,2,2-pentadeuterioethyl)acetamide |
| Molecular Weight (g/mol) | 310.36 |
| Formula Weight | 310.159 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction