Learn More
(±)-Warfarin-d5 (phenyl-d5), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | (±)-Warfarin D5 (phenyl-D5) |
| CAS | 75472-93-4 |
| Molecular Formula | C19 2H5 H11 O4 |
| Quantity | 0.01 g |
| Synonym | (±)-Warfarin D5 (phenyl-D5), 2 H-1-Benzopyran-2-one, 4-hydroxy-3-[3-oxo-1-(phenyl-d5)butyl]- (9 CI), (±)-4-Hydroxy-3-[3-oxo-1-(phenyl-d5)butyl]-2 H-1-benzopyran-2-one |
| InChI Formula | InChI=1 S/C19H16O4/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22/h2-10,15,21 H,11H2,1H3/i2D,3 D,4 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(CC(=O)C)C2=C(O)c3ccccc3OC2=O)c([2 H])c1[2 H] |
| IUPAC Name | 4-hydroxy-3-[3-oxo-1-(2,3,4,5,6-pentadeuteriophenyl)butyl]chromen-2-one |
| Molecular Weight (g/mol) | 313.36 |
| Formula Weight | 313.1362 g/mol |
| Show More |