missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vortioxetine-d8 HBr (piperazine-d8), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Vortioxetine Hydrobromide-D8 |
| Molecular Formula | C18 D8 H14 N2 S . Br H |
| Quantity | 0.005 g |
| Synonym | 1-[2-[(2,4-Dimethylphenyl)thio]phenyl]piperazine-2,2,3,3,5,5,6,6-d8 hydrobromide, 1-[2-(2,4-Dimethylphenylsulfanyl)phenyl]piperazine-2,2,3,3,5,5,6,6-d8 hydrobromide, Lu AA 21004-d8 hydrobromide, Vortioxetine-d8 hydrobromide, Vortioxetine-D8 (piperazine-2,2,3,3,5,5,6,6-D8) hydrobromide |
| InChI Formula | InChI=1 S/C18H22N2S.BrH/c1-14-7-8-17(15(2)13-14)21-18-6-4-3-5-16(18)20-11-9-19-10-12-20;/h3-8,13,19 H,9-12H2,1-2H3;1 H/i9D2,10D2,11D2,12D2; |
| SMILES | Br.[2 H]C1([2 H])NC([2 H])([2 H])C([2 H])([2 H])N(c2ccccc2Sc3ccc(C)cc3C)C1([2 H])[2 H] |
| IUPAC Name | 2,2,3,3,5,5,6,6-octadeuterio-1-[2-(2,4-dimethylphenyl)sulfanylphenyl]piperazine;hydrobromide |
| Molecular Weight (g/mol) | 387.407 |
| Formula Weight | 386.127 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction