Learn More
Valdecoxib-d3 (methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Valdecoxib methyl-d3 |
| CAS | 1219794-90-7 |
| Molecular Formula | C16 2H3 H11 N2 O3 S |
| Quantity | 0.005 g |
| InChI Formula | InChI=1 S/C16H14N2O3S/c1-11-15(12-7-9-14(10-8-12)22(17,19)20)16(18-21-11)13-5-3-2-4-6-13/h2-10 H,1H3,(H2,17,19,20)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])c1onc(c2ccccc2)c1c3ccc(cc3)S(=O)(=O)N |
| IUPAC Name | 4-[3-phenyl-5-(trideuteriomethyl)-1,2-oxazol-4-yl]benzenesulfonamide |
| Molecular Weight (g/mol) | 317.38 |
| Formula Weight | 317.0913 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |