missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Triadimefon-d4 (4-chlorophenoxy-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Triadimefon D4 (phenoxy D4) |
| Molecular Formula | C14 D4 H12 Cl N3 O2 |
| Quantity | 0.005 g |
| Synonym | Triadimefon D4 (phenoxy D4), 1-(4-Chlorophenoxy-2,3,5,6-d4)-3,3-dimethyl-1-(1 H-1,2,4-triazol-1-yl)-2-butanone, Triadimefon-d4, (±)-Triadimefon-d4 (4-chlorophenoxy-d4), Triadimefon-D4 (4-chlorophenoxy-D4) |
| InChI Formula | InChI=1 S/C14H16ClN3O2/c1-14(2,3)12(19)13(18-9-16-8-17-18)20-11-6-4-10(15)5-7-11/h4-9,13 H,1-3H3/i4D,5 D,6 D,7 D |
| SMILES | [2 H]c1c([2 H])c(OC(C(=O)C(C)(C)C)n2cncn2)c([2 H])c([2 H])c1Cl |
| IUPAC Name | 1-(4-chloro-2,3,5,6-tetradeuteriophenoxy)-3,3-dimethyl-1-(1,2,4-triazol-1-yl)butan-2-one |
| Molecular Weight (g/mol) | 297.773 |
| Formula Weight | 297.118 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction