Learn More
Testosterone-2,2,4,6,6-d5 Propionate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Specifications
Specifications
| Chemical Name or Material | Testosterone-2,2,4,6,6-D5 Propionate |
| Molecular Formula | C22 D5 H27 O3 |
| Quantity | 0.01 g |
| Synonym | Testosterone-2,2,4,6,6-d5 17-propanoate, Testosterone-d5 propionate, Testosterone-2,2,4,6,6-d5 propionate, Testosterone-2,2,4,6,6-D5 propionate, (17β)-17-(1-Oxopropoxy)androst-4-en-3-one-2,2,4,6,6-d5 |
| InChI Formula | InChI=1 S/C22H32O3/c1-4-20(24)25-19-8-7-17-16-6-5-14-13-15(23)9-11-21(14,2)18(16)10-12-22(17,19)3/h13,16-19 H,4-12H2,1-3H3/t16-,17-,18-,19-,21-,22-/m0/s1/i5D2,9D2,13 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C[C@H]3 [C@@H]4 CC[C@H](OC(=O)CC)[C@@]4(C)CC[C@@H]3 [C@@]2(C)CC([2 H])([2 H])C1=O |
| IUPAC Name | [(8 R,9 S,10 R,13 S,14 S,17 S)-2,2,4,6,6-pentadeuterio-10,13-dimethyl-3-oxo-1,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] propanoate |
| Molecular Weight (g/mol) | 349.519 |
| Formula Weight | 349.267 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |