missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone-2,2,4,6,6-d5 Acetate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Specifications
Specifications
| Chemical Name or Material | Testosterone-2,2,4,6,6-D5 Acetate |
| Molecular Formula | C21 D5 H25 O3 |
| Quantity | 0.01 g |
| Synonym | 2,2,4,6,6-Pentadeuterio-4-androsten-17β-ol-3-one 17-acetate, 17-O-Acetyltestosterone-2,2,4,6,6-d5, (17β)-17-Acetoxyandrost-4-ene-3-one-2,2,4,6,6-d5, Testosterone-2,2,4,6,6-d5 17-acetate, Testosterone-2,2,4,6,6-d5 acetate, Testosterone-2,2,4,6,6-D5 acetate |
| InChI Formula | InChI=1 S/C21H30O3/c1-13(22)24-19-7-6-17-16-5-4-14-12-15(23)8-10-20(14,2)18(16)9-11-21(17,19)3/h12,16-19 H,4-11H2,1-3H3/t16-,17-,18-,19-,20-,21-/m0/s1/i4D2,8D2,12 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C[C@H]3 [C@@H]4 CC[C@H](OC(=O)C)[C@@]4(C)CC[C@@H]3 [C@@]2(C)CC([2 H])([2 H])C1=O |
| IUPAC Name | [(8 R,9 S,10 R,13 S,14 S,17 S)-2,2,4,6,6-pentadeuterio-10,13-dimethyl-3-oxo-1,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] acetate |
| Molecular Weight (g/mol) | 335.492 |
| Formula Weight | 335.251 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction