missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamerazine-d4 (benzene-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Sulfamerazine-d4 |
| CAS | 1020719-84-9 |
| Molecular Formula | C11 2H4 H8 N4 O2 S |
| Quantity | 0.01 g |
| Synonym | Benzene-2,3,5,6-d4-sulfonamide, 4-amino-N-(4-methyl-2-pyrimidinyl)-, Sulfamerazine D4 |
| InChI Formula | InChI=1 S/C11H12N4O2S/c1-8-6-7-13-11(14-8)15-18(16,17)10-4-2-9(12)3-5-10/h2-7 H,12H2,1H3,(H,13,14,15)/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1N)S(=O)(=O)Nc2nccc(C)n2 |
| IUPAC Name | 4-amino-2,3,5,6-tetradeuterio-N-(4-methylpyrimidin-2-yl)benzenesulfonamide |
| Molecular Weight (g/mol) | 268.33 |
| Formula Weight | 268.0932 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction