missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfadoxine-d4 (benzene-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Sulfadoxine (Phenylene-d4) |
| CAS | 1330266-05-1 |
| Molecular Formula | C12 D4 H10 N4 O4 S |
| Quantity | 0.01 g |
| Synonym | 4-Amino-2,3,5,6-tetradeuterio-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulfonamide, 4-Amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulfonamide-2,3,5,6-d4, Sulfadoxine-d4, Sulfadoxine-d4 (benzene-d4), Sulfadoxine-D4 (phenylene-D4) |
| InChI Formula | InChI=1 S/C12H14N4O4S/c1-19-10-11(14-7-15-12(10)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7 H,13H2,1-2H3,(H,14,15,16)/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1N)S(=O)(=O)Nc2ncnc(OC)c2OC |
| IUPAC Name | 4-amino-2,3,5,6-tetradeuterio-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulfonamide |
| Molecular Weight (g/mol) | 314.354 |
| Formula Weight | 314.099 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction