missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stavudine-alpha,alpha,alpha,6-d4, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Stavudine-D4 (thymine-alpha,alpha,alpha,6-D4) |
| CAS | 1219803-67-4 |
| Molecular Formula | C10 D4 H8 N2 O4 |
| Quantity | 0.005 g |
| Synonym | Stavudine-d4, Stavudine-α,α,α,6-d4, Stavudine-D4 (thymine-α,α,α,6-D4), 1-[(2 R,5 S)-5-(Hydroxymethyl)-2,5-dihydrofuran-2-yl)-5-(methyl-d3)pyrimidine-2,4(1 H,3 H)-dione-6-d, 1-(2,3-Dideoxy-β-D-glycero-pent-2-enofuranosyl)thymine-α,α,α,6-d4, 3'-Deoxy-2',3'-didehydrothymidine-d4 |
| InChI Formula | InChI=1 S/C10H12N2O4/c1-6-4-12(10(15)11-9(6)14)8-3-2-7(5-13)16-8/h2-4,7-8,13 H,5H2,1H3,(H,11,14,15)/t7-,8+/m0/s1/i1D3,4 D |
| SMILES | [2 H]C1=C(C(=O)NC(=O)N1[C@@H]2 O[C@H](CO)C=C2)C([2 H])([2 H])[2 H] |
| IUPAC Name | 6-deuterio-1-[(2 R,5 S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-(trideuteriomethyl)pyrimidine-2,4-dione |
| Molecular Weight (g/mol) | 228.238 |
| Formula Weight | 228.105 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction