missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Pregnenolone-17alpha,21,21,21-d4 Sulfate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Pregnenolone-17 alpha,21,21,21-D4 Sulfate Sodium Salt |
| CAS | 1485492-21-4 |
| Molecular Formula | C21H27D4NaO5S |
| Quantity | 0.005 g |
| Synonym | (3β)-3-(Sulfooxy)pregn-5-en-20-one-17α,21,21,21-d4 sodium salt, 3β-Hydroxypregn-5-en-20-one-17α,21,21,21-d4 hydrogen sulfate sodium salt, Pregnenolone-17α,21,21,21-D4 sulfate sodium salt, Pregnenolone-3-sulfate-17α,21,21,21-d4 sodium salt, Pregnenolone-17α,21,21,21-d4 3β-sulfate sodium salt, Sodium Pregnenolone-17 alpha,21,21,21-d4 Sulfate, Sodium Pregnenolone-17 alpha,21,21,21-d4 Sulfate, Sodium Pregnenolone-17 alpha,21,21,21-d4 Sulfate, Sodium Pregnenolone-17 alpha,21,21,21-d4 Sulfate |
| InChI Formula | InChI=1 S/C21H32O5S.Na/c1-13(22)17-6-7-18-16-5-4-14-12-15(26-27(23,24)25)8-10-20(14,2)19(16)9-11-21(17,18)3;/h4,15-19 H,5-12H2,1-3H3,(H,23,24,25);/q;+1/p-1/t15-,16-,17+,18-,19-,20-,21+;/m0./s1/i1D3,17 D; |
| SMILES | C[C@@](C1=CC2)(CC[C@H](O[S](=O)(O[Na])=O)C1)[C@]3([H])[C@]2([H])[C@@](CC[C@]4([2 H])C(C([2 H])([2 H])[2 H])=O)([H])[C@]4(C)CC3 |
| IUPAC Name | sodium (3 S,8 S,9 S,10 R,13 S,14 S,17 S)-17-(acetyl-d3)-10,13-dimethyl-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1 H-cyclopenta[a]phenanthren-3-yl-17-d sulfate |
| Molecular Weight (g/mol) | 422.55 |
| Formula Weight | 422.2 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction