missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dehydroepiandrosterone-16,16-d2 Sulfate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Sodium Prasterone-16,16-D2 Sulfate |
| CAS | 352431-52-8 |
| Molecular Formula | C19 D2 H25 O5 S . Na |
| Quantity | 0.01 g |
| Synonym | Androst-5-en-17-one-16,16-d2, 3-(sulfooxy)-, sodium salt, (3β)- (9 CI), (3β)-3-(Sulfooxy)androst-5-en-17-one-16,16-d2 sodium salt, Dehydroepiandrosterone-16,16-d2 sulfate sodium salt, Prasterone-16,16-d2 sodium sulfate, Sodium dehydroepiandrosterone-16,16-d2 sulfate, Sodium dehydroepiandrosterone-3-sulfate-16,16-d2, Sodium prasterone-16,16-d2 sulfate |
| InChI Formula | InChI=1 S/C19H28O5S.Na/c1-18-9-7-13(24-25(21,22)23)11-12(18)3-4-14-15-5-6-17(20)19(15,2)10-8-16(14)18;/h3,13-16 H,4-11H2,1-2H3,(H,21,22,23);/q;+1/p-1/t13-,14-,15-,16-,18-,19-;/m0./s1/i6D2; |
| SMILES | [Na+].[2 H]C1([2 H])C[C@H]2 [C@@H]3 CC=C4C[C@H](CC[C@]4(C)[C@H]3 CC[C@]2(C)C1=O)OS(=O)(=O)[O-] |
| IUPAC Name | sodium;[(3 S,8 R,9 S,10 R,13 S,14 S)-16,16-dideuterio-10,13-dimethyl-17-oxo-2,3,4,7,8,9,11,12,14,15-decahydro-1 H-cyclopenta[a]phenanthren-3-yl] sulfate |
| Molecular Weight (g/mol) | 392.4818 |
| Formula Weight | 392.1602 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction