missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Cholesterol-25,26,26,26,27,27,27-d7 Sulfate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Sodium Cholesterol-25,26,26,26,27,27,27-D7 Sulfate |
| CAS | 2687961-01-7 |
| Molecular Formula | C27H38D7NaO4S |
| Quantity | 0.005 g |
| Synonym | Cholest-5-en-25,26,26,26,27,27,27-d7-3-ol, 3-(hydrogen sulfate), sodium salt (1:1), (3β)- (ACI), Cholesteryl-25,26,26,26,27,27,27-d7 sodium sulfate, Cholesterol-25,26,26,26,27,27,27-d7 3-sulfate sodium salt, Cholesteryl-25,26,26,26,27,27,27-d7 sulfate sodium salt, Sodium cholesteryl-25,26,26,26,27,27,27-d7 sulfate, (3β)-Cholest-5-en-3-ol-25,26,26,26,27,27,27-d7 3-(hydrogen sulfate) sodium salt |
| InChI Formula | InChI=1 S/C27H46O4S.Na/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(31-32(28,29)30)13-15-26(20,4)25(22)14-16-27(23,24)5;/h9,18-19,21-25 H,6-8,10-17H2,1-5H3,(H,28,29,30);/q;+1/p-1/t19-,21+,22+,23-,24+,25+,26+,27-;/m1./s1/i1D3,2D3,18 D; |
| SMILES | C[C@]12 C(C[C@H](CC2)OS(=O)(O[Na])=O)=CC[C@]3([H])[C@]1([H])CC[C@]4([C@@]3([H])CC[C@]4([H])[C@H](C)CCCC([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C |
| IUPAC Name | sodium (3 S,8 S,9 S,10 R,13 R,14 S,17 R)-10,13-dimethyl-17-((R)-6-(methyl-d3)heptan-2-yl-6,7,7,7-d4)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1 H-cyclopenta[a]phenanthren-3-yl sulfate |
| Molecular Weight (g/mol) | 495.75 |
| Formula Weight | 495.34 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction