Learn More
Rimantadine-d4 HCl (ethyl-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Rimantadine-d4 Hydrochloride |
| CAS | 350818-67-6 |
| Molecular Formula | C12 D4 H17 N . Cl H |
| Quantity | 0.01 g |
| Synonym | Tricyclo[3.3.1.13,7 ]decane-1-methan-α-d-amine, α-(methyl-d3)-, hydrochloride (9 CI), α-(Methyl-d3)tricyclo[3.3.1.13,7 ]decane-1-methan-α-d-amine hydrochloride, (±)-Rimantadine-d4 hydrochloride, 1-(1-Adamantyl)ethylamine-1,2,2,2-d4 hydrochloride, 1-(1-Aminoethyl-1,2,2,2-d4)adamantane hydrochloride, 1-(Adamantan-1-yl)ethanamine-d4 hydrochloride, 1-Rimantadine-d4 hydrochloride, Rimantadine-d4 hydrochloride, Rimantadine-D4 (aminoethyl-1,2,2,2-D4) hydrochloride |
| InChI Formula | InChI=1 S/C12H21N.ClH/c1-8(13)12-5-9-2-10(6-12)4-11(3-9)7-12;/h8-11 H,2-7,13H2,1H3;1 H/i1D3,8 D; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])(N)C12CC3CC(CC(C3)C1)C2 |
| IUPAC Name | 1-(1-adamantyl)-1,2,2,2-tetradeuterioethanamine;hydrochloride |
| Molecular Weight (g/mol) | 219.787 |
| Formula Weight | 219.169 g/mol |
| Show More |