Learn More
(±)-Propafenone-d7 HCl (n-propyl-d7), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Propafenone-D7 (propyl-D7) Hydrochloride |
| CAS | 1219799-06-0 |
| Molecular Formula | C21 D7 H20 N O3 . Cl H |
| Quantity | 0.01 g |
| Synonym | 1-[2-[2-Hydroxy-3-(heptadeuteriopropylamino)propoxy]phenyl]-3-phenylpropan-1-one hydrochloride, Propafenone-d7 hydrochloride, Propafenone-D7 (propyl-D7) hydrochloride, 1-[2-[2-Hydroxy-3-(propyl-d7-amino)propoxy]phenyl]-3-phenyl-1-propanone hydrochloride |
| InChI Formula | InChI=1 S/C21H27NO3.ClH/c1-2-14-22-15-18(23)16-25-21-11-7-6-10-19(21)20(24)13-12-17-8-4-3-5-9-17;/h3-11,18,22-23 H,2,12-16H2,1H3;1 H/i1D3,2D2,14D2; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])NCC(O)COc1ccccc1C(=O)CCc2ccccc2 |
| IUPAC Name | 1-[2-[3-(1,1,2,2,3,3,3-heptadeuteriopropylamino)-2-hydroxypropoxy]phenyl]-3-phenylpropan-1-one;hydrochloride |
| Molecular Weight (g/mol) | 384.948 |
| Formula Weight | 384.22 g/mol |
| Show More |