missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pheniramine-d6 Maleate (N,N-dimethyl-d6), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Pheniramine-D6 (N,N-dimethyl-D6) Maleate |
| Molecular Formula | C16 D6 H14 N2 . C4 H4 O4 |
| Quantity | 0.005 g |
| Synonym | N,N-Di(methyl-d3)-γ-phenyl-2-pyridinepropanamine (Z)-2-butenedioate (1:1), (3 RS)-N,N-Di(methyl-d3)-3-phenyl-3-(pyridin-2-yl)propan-1-amine maleate, Pheniramine-d6 maleate, Pheniramine-D6 (N,N-dimethyl-D6) maleate |
| InChI Formula | InChI=1 S/C16H20N2.C4H4O4/c1-18(2)13-11-15(14-8-4-3-5-9-14)16-10-6-7-12-17-16;5-3(6)1-2-4(7)8/h3-10,12,15 H,11,13H2,1-2H3;1-2 H,(H,5,6)(H,7,8)/b;2-1-/i1D3,2D3; |
| SMILES | [2 H]C([2 H])([2 H])N(CCC(c1ccccc1)c2ccccn2)C([2 H])([2 H])[2 H].OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;3-phenyl-3-pyridin-2-yl-N,N-bis(trideuteriomethyl)propan-1-amine |
| Molecular Weight (g/mol) | 362.452 |
| Formula Weight | 362.211 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction