Learn More
Phenelzine-d5 Sulfate (phenyl-d5), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Phenelzine-D5 (phenyl-D5) Sulfate |
| CAS | 1219798-40-9 |
| Molecular Formula | C8 D5 H7 N2 . H2 O4 S |
| Quantity | 0.01 g |
| Synonym | [2-(Phenyl-d5)ethyl]hydrazine sulfate, 1-(2-Phenethyl)hydrazine-d5 sulfate, 1-Hydrazino-2-(phenyl-d5)ethane sulfate, Phenelzine-d5 sulfate, Phenelzine-D5 (phenyl-D5) sulfate |
| InChI Formula | InChI=1 S/C8H12N2.H2O4S/c9-10-7-6-8-4-2-1-3-5-8;1-5(2,3)4/h1-5,10 H,6-7,9H2;(H2,1,2,3,4)/i1D,2 D,3 D,4 D,5 D; |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CCNN)c([2 H])c1[2 H].OS(=O)(=O)O |
| IUPAC Name | 2-(2,3,4,5,6-pentadeuteriophenyl)ethylhydrazine;sulfuric acid |
| Molecular Weight (g/mol) | 239.304 |
| Formula Weight | 239.099 g/mol |
| Show More |