missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Penicillin V-d5 Potassium Salt (phenyl-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Phenoxymethylpenicillin-D5 (phenyl-D5) Potassium |
| CAS | 2699607-22-0 |
| Molecular Formula | C16 D5 H12 N2 O5 S . K |
| Quantity | 0.005 g |
| Synonym | (2 S,5 R,6 R)-3,3-Dimethyl-7-oxo-6-[[2-(phenoxy-2,3,4,5,6-d5)acetyl]amino]-4-thia-1-azabicyclo[3.2.0 ]heptane-2-carboxylic acid potassium salt, (2 S,5 R,6 R)-3,3-Dimethyl-7-oxo-6-[[2-(2,3,4,5,6-pentadeuteriophenoxy)acetyl]amino]-4-thia-1-azabicyclo[3.2.0 ]heptane-2-carboxylic acid potassium salt, (2 S,5 R,6 R)-3,3-Dimethyl-7-oxo-6-[[(phenoxy-d5)acetyl]amino]-4-thia-1-azabicyclo[3.2.0 ]heptane-2-carboxylic acid potassium salt, Penicillin V-d5 potassium salt, Phenoxymethylpenicillin-d5 potassium salt, Phenoxymethylpenicillin-D5 (phenyl-D5) potassium salt, Penicillin V-d5 potassium, Phenoxymethylpenicillin-d5 potassium, Phenoxymethylpenicillin-D5 (phenyl-D5) potassium |
| InChI Formula | InChI=1 S/C16H18N2O5S.K/c1-16(2)12(15(21)22)18-13(20)11(14(18)24-16)17-10(19)8-23-9-6-4-3-5-7-9;/h3-7,11-12,14 H,8H2,1-2H3,(H,17,19)(H,21,22);/q;+1/p-1/t11-,12+,14-;/m1./s1/i3D,4 D,5 D,6 D,7 D; |
| SMILES | [K+].[2 H]c1c([2 H])c([2 H])c(OCC(=O)N[C@H]2 [C@H]3 SC(C)(C)[C@@H](N3C2=O)C(=O)[O-])c([2 H])c1[2 H] |
| IUPAC Name | potassium;(2 S,5 R,6 R)-3,3-dimethyl-7-oxo-6-[[2-(2,3,4,5,6-pentadeuteriophenoxy)acetyl]amino]-4-thia-1-azabicyclo[3.2.0 ]heptane-2-carboxylate |
| Molecular Weight (g/mol) | 393.511 |
| Formula Weight | 393.081 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction