missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olanzapine-d3 (4-methyl-d3-piperazinyl), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Olanzapine-methyl-D3 |
| CAS | 786686-79-1 |
| Molecular Formula | C17 2H3 H17 N4 S |
| Quantity | 0.005 g |
| Synonym | 2-Methyl-4-(4-methyl-d3-1-piperazinyl)-10 H-thieno[2,3-b][1,5 ]benzodiazepine, Integrol-d3, LY 170053-d3, Lanzac-d3, Oleanz-d3, 2-Methyl-4-[4-(methyl-d3)-1-piperazinyl]-10 H-thieno[2,3-b][1,5 ]benzodiazepine (9 CI), 2-Methyl-4-[4-(methyl-d3)-1-piperazinyl]-10 H-thieno[2,3-b][1,5 ]benzodiazepine (9 CI), 2-Methyl-4-[4-(methyl-d3)-1-piperazinyl]-10 H-thieno[2,3-b][1,5 ]benzodiazepine (9 CI), 2-Methyl-4-[4-(methyl-d3)-1-piperazinyl]-10 H-thieno[2,3-b][1,5 ]benzodiazepine (9 CI), 2-Methyl-4-[4-(methyl-d3)-1-piperazinyl]-10 H-thieno[2,3-b][1,5 ]benzodiazepine (9 CI) |
| InChI Formula | InChI=1 S/C17H20N4S/c1-12-11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)22-12/h3-6,11,19 H,7-10H2,1-2H3/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])N1CCN(CC1)C2=Nc3ccccc3Nc4sc(C)cc24 |
| IUPAC Name | 2-methyl-4-[4-(trideuteriomethyl)piperazin-1-yl]-10 H-thieno[2,3-b][1,5 ]benzodiazepine |
| Molecular Weight (g/mol) | 315.45 |
| Formula Weight | 315.1597 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction