Learn More
Octanoyl-L-carnitine-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Octanoyl L-Carnitine-d3 Chloride |
| CAS | 1334532-24-9 |
| Molecular Formula | C15 D3 H27 N O4 . Cl |
| Quantity | 0.01 g |
| Synonym | Octanoyl-L-carnitine-d3 HCl (N-methyl-d3) |
| InChI Formula | InChI=1 S/C15H29NO4.ClH/c1-5-6-7-8-9-10-15(19)20-13(11-14(17)18)12-16(2,3)4;/h13H,5-12H2,1-4H3;1 H/t13-;/m1./s1/i2D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCCCC |
| IUPAC Name | [(2 R)-3-carboxy-2-octanoyloxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 326.87 |
| Formula Weight | 326.2052 g/mol |
| Show More |